Learn More
Cayman Chemical 19-hydroxy Cholesterol, Purity: ≥98%, CAS: 561-63-7, 10 mg
SDP
Catalog No. 502972817
Encompass_Preferred
An oxysterol formed during the metabolic oxidation of cholesterol; used as an internal standard for the quantitative determination of sterols by mass spectroscopic analysis
Catalog No. 50-297-2817 Supplier Cayman Chemical Supplier No. 1520910 MG
An oxysterol formed during the metabolic oxidation of cholesterol; used as an internal standard for the quantitative determination of sterols by mass spectroscopic analysis