Learn More
Cayman Chemical 19-hydroxy Cholesterol, Purity: ≥98%, CAS: 561-63-7, 5 mg
SDP
Catalog No. 502972079
Encompass_Preferred
An oxysterol formed during the metabolic oxidation of cholesterol; used as an internal standard for the quantitative determination of sterols by mass spectroscopic analysis
Catalog No. 50-297-2079 Supplier Cayman Chemical Supplier No. 152095 MG
An oxysterol formed during the metabolic oxidation of cholesterol; used as an internal standard for the quantitative determination of sterols by mass spectroscopic analysis