Learn More
Medchemexpress LLC HY-N7019 5mg Medchemexpress, 19-Hydroxybufalin CAS:39844-86-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN70195MG
Encompass_Preferred
Medchemexpress, HY-N7019 5mg 19-Hydroxybufalin CAS:39844-86-5 19-Hydroxybufalin is a bufadienolide, inhibits epithelial-mesenchymal transition and attenuates the migration and invasion of PC3 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2979