Learn More
Cayman Chemical 19-Noretiocholanolone, Purity: ≥98%, CAS: 33036-33-8, 10 mg
SDP
Catalog No. 502964550
Encompass_Preferred
An analytical reference standard categorized as an anabolic androgenic steroid metabolite of nandrolone; intended for research and forensic applications
Catalog No. 50-296-4550 Supplier Cayman Chemical Supplier No. 2268410 MG
An analytical reference standard categorized as an anabolic androgenic steroid metabolite of nandrolone; intended for research and forensic applications