Learn More
Medchemexpress LLC 19-Norpregn-4-en-20-yn-3-one, 17-[(1-oxoheptyl)oxy]-, (17α)- | 3836-23-5 | 99.5% | C27H38O3 | 500 MG
SDP
Catalog No. 5000432979
Encompass_Preferred
Norethisterone enanthate is a long-acting parenteral progestogen. It is orally active and functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. This product is for research use only and not sold to patients.
  • Long-acting parenteral progestogen
  • Orally active
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can be used as an intramuscular contraceptive agent (in vitro)
  • Inhibits fertility in female rats (in vivo)
Catalog No. 50-004-32979 Supplier Medchemexpress LLC Supplier No. HYA0285500MG
Norethisterone enanthate is a long-acting parenteral progestogen. It is orally active and functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules possessing Azide groups. This product is for research use only and not sold to patients.
  • Long-acting parenteral progestogen
  • Orally active
  • Functions as a click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can be used as an intramuscular contraceptive agent (in vitro)
  • Inhibits fertility in female rats (in vivo)