Learn More
AdipoGen 19-O-Acetylchaetoglobosin A
SDP
Supplier:  AdipoGen BVT0383M005
Encompass_Preferred
Chemical. CAS 50939-69-0. Formula C34H38N2O6. MW 570.7. Isolated from Chaetomium sp. Cytochalasan alkaloid, derivative of chaetoglobosin A Prod. No. BVT-0092 http//www.adipogen.com/bvt-0092/chaetoglobosin-a.html . Mycotoxin. Phytotoxin.
Catalog No. 50-256-1957