Learn More
Cayman Chemical 19 RHETE Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1000776725 UG
Encompass_Preferred
A potent vasodilator of renal arterioles; 30% of the metabolite composition of arachidonic acid by CYP2E1; potent vasodilator of renal preglomerular vessels; blocks 20-HETE-induced vasoconstriction of renal arterioles at 1 µM
Catalog No. 50-288-8020