Learn More
Cayman Chemical 19 SHETE Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1000776625 UG
Encompass_Preferred
Potent vasodilator of renal preglomerular vessels; 70% of the metabolite composition of arachidonic acid by CYP2E1; stimulates both renal sodium-potassium ATPase and volume absorption in the rabbit proximal straight tubule
Catalog No. 50-288-7658