Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 19-(Z)-Akuammidine | 113973-31-2 | 352.43 | 1 MG
SDP

Catalog No. 5000416157
Encompass_Preferred

(Z)-Akuammidine, also known as 19-(Z)-Akuammidine or (Z)-Rhazine, is a natural compound isolated from Gelsemium elegans. This solid, white to off-white compound boasts a purity of 95.0% and is intended exclusively for research applications.

  • Isolated from Gelsemium elegans.
  • Solid, white to off-white appearance.
  • Purity of 95.0%.
  • Designed for research use only.
  • Optimal storage at -20°C, protected from light.
  • For solutions, store at -80°C for up to 6 months or -20°C for 1 month, protected from light.

Catalog No. 50-004-16157 Supplier Medchemexpress LLC Supplier No. HYN09691MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(Z)-Akuammidine, also known as 19-(Z)-Akuammidine or (Z)-Rhazine, is a natural compound isolated from Gelsemium elegans. This solid, white to off-white compound boasts a purity of 95.0% and is intended exclusively for research applications.

  • Isolated from Gelsemium elegans.
  • Solid, white to off-white appearance.
  • Purity of 95.0%.
  • Designed for research use only.
  • Optimal storage at -20°C, protected from light.
  • For solutions, store at -80°C for up to 6 months or -20°C for 1 month, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.