Learn More
Cole-Parmer Cylindrical Dewar Flask with Handle, 1900 mL
Supplier:  Cole-Parmer 0377111
Encompass_Preferred
These Dewars feature an exterior metal housing with a boroscilicate glass interior. There are no pads between walls to eliminate heat transfer potential. These are ideal for temporary storage or for transfer applications. Dewar flasks come with lids.
Catalog No. NC0632710