Learn More
Chemglass Life Sciences Dewar Flask, Wide Mouth, Metal Housing, Carrying Handle, 1900mL Capacity, 190mm ID X 120mm Inside Depth, 150mm OD X 252mm Height
SDP
Catalog No. CG159502
Encompass_Preferred
Dewar Flask, Wide Mouth, Metal Housing, Carrying Handle, 1900mL Capacity, 190mm ID X 120mm Inside Depth, 150mm OD X 252mm Height. Similar to CG-1594, but with the addition of a convenient carrying handle. Supplied complete with a vented polyethylene stopper to minimize evaporation. Exterior dimensions listed do not include carrying handle.
Catalog No. CG159502 Supplier Chemglass Life Sciences Supplier No. CG159502
Dewar Flask, Wide Mouth, Metal Housing, Carrying Handle, 1900mL Capacity, 190mm ID X 120mm Inside Depth, 150mm OD X 252mm Height. Similar to CG-1594, but with the addition of a convenient carrying handle. Supplied complete with a vented polyethylene stopper to minimize evaporation. Exterior dimensions listed do not include carrying handle.