Learn More
Chemglass Life Sciences Dewar Flask, Wide Mouth, Metal Housing, 1900mL Capacity, 120mm ID X 190mm Inside Depth, 150mm OD X 252mm Height
SDP
Catalog No. CG159404
Encompass_Preferred
Dewar Flask, Wide Mouth, Metal Housing, 1900mL Capacity, 120mm ID X 190mm Inside Depth, 150mm OD X 252mm Height. Similar to CG-1593, but with a full length aluminum housing which extends to the top rim for added protection. Supplied complete with a vented polyethylene stopper to minimize evaporation.
Catalog No. CG159404 Supplier Chemglass Life Sciences Supplier No. CG159404
Dewar Flask, Wide Mouth, Metal Housing, 1900mL Capacity, 120mm ID X 190mm Inside Depth, 150mm OD X 252mm Height. Similar to CG-1593, but with a full length aluminum housing which extends to the top rim for added protection. Supplied complete with a vented polyethylene stopper to minimize evaporation.