Learn More
Chemglass Life Sciences Dewar Flask, Cylindrical, Wide Mouth, 1900mL Capacity, 120mm ID X 190mm Inside Depth, 150mm OD X 252mm Height
SDP
Catalog No. CG159304
Encompass_Preferred
Dewar Flask, Cylindrical, Wide Mouth, 1900mL Capacity, 120mm ID X 190mm Inside Depth, 150mm OD X 252mm Height. Dewar flask with weighted metal base for stability and protective plastic mesh cover providing a firm grip and a cushion in the event the flask is bumped. Supplied complete with a vented polyethylene stopper to minimize evaporation.
Catalog No. CG159304 Supplier Chemglass Life Sciences Supplier No. CG159304
Dewar Flask, Cylindrical, Wide Mouth, 1900mL Capacity, 120mm ID X 190mm Inside Depth, 150mm OD X 252mm Height. Dewar flask with weighted metal base for stability and protective plastic mesh cover providing a firm grip and a cushion in the event the flask is bumped. Supplied complete with a vented polyethylene stopper to minimize evaporation.