Learn More
MSC Howard Leight Earplugs, 30 dB NRR, Foam, Contoured, Uncorded, 200/Pk.
Catalog No. 502370294
Encompass
These disposable earplugs feature a contoured shape for a comfortable fit and a Noise Reduction Rating (NRR) of 30 dB. Made from green foam, they are uncorded and designed for roll-down insertion. The earplugs are universal in size and supplied in a package of 200 pairs.
  • Disposable
  • Uncorded attachment style
  • Contoured plug shape
  • Noise reduction rating of 30 dB
  • Green plug color
  • Foam plug material
  • Roll down insertion method
  • Universal size
  • Package quantity of 200 pairs
  • Non-metal detectable
  • Maximum Lite series
Catalog No. 50-237-0294 Supplier MSC Supplier No. 09798026
These disposable earplugs feature a contoured shape for a comfortable fit and a Noise Reduction Rating (NRR) of 30 dB. Made from green foam, they are uncorded and designed for roll-down insertion. The earplugs are universal in size and supplied in a package of 200 pairs.
  • Disposable
  • Uncorded attachment style
  • Contoured plug shape
  • Noise reduction rating of 30 dB
  • Green plug color
  • Foam plug material
  • Roll down insertion method
  • Universal size
  • Package quantity of 200 pairs
  • Non-metal detectable
  • Maximum Lite series