Learn More
Ace Glass, Inc. Porcelain desiccator plate, 190mm, 5mm holes, no feet, cs/3
SDP
Catalog No. 50983480
Encompass
190mm porcelain desiccator plate with many 5mm holes and one large center hole. Plate is glazed on one side and does not have feet. Plate can withstand temperature up to 1150 degrees C.
Catalog No. 50-983-480 Supplier Ace Glass, Inc. Supplier No. 1325404
190mm porcelain desiccator plate with many 5mm holes and one large center hole. Plate is glazed on one side and does not have feet. Plate can withstand temperature up to 1150 degrees C.