Learn More
Alkali Scientific 190uL Beckman Biomek FX/NX Mul
SDP
Catalog No. 503028866
Encompass_Preferred
Alkali Scientificdesigns its P250 black conductive automation tips to ensure optimal compatibility with Beckman Coulter Biomek i-Series (i5 and i7) and previous NX/FX/FXP robotic liquid handlers These tips require no additional labware definition and existing automation control software protocols remain unchanged The Alkali ScientificP250 (250 L) tip type is one of the most popular choices in the 96-format category Known for its excellent sub-microliter performance (precision and accuracy) this tip offers versatility in protocol design due to its volume capacity and length It is compatible with all 96 multi-channel heads on Biomek i-Series (i5 and i7) NX/FX/FXP and Multimek Alkali Scientificmanufactures its P250 black conductive tips from premium virgin polypropylene and utilizes advanced polishing techniques to ensure minimal liquid retention without using Specifications Working volume 190 00 L Well format 96 96 384 plate accessible Head compatibility 96 multi-channel Beckman
Catalog No. 50-302-8866 Supplier Alkali Scientific Supplier No. FXBS180
Alkali Scientificdesigns its P250 black conductive automation tips to ensure optimal compatibility with Beckman Coulter Biomek i-Series (i5 and i7) and previous NX/FX/FXP robotic liquid handlers These tips require no additional labware definition and existing automation control software protocols remain unchanged The Alkali ScientificP250 (250 L) tip type is one of the most popular choices in the 96-format category Known for its excellent sub-microliter performance (precision and accuracy) this tip offers versatility in protocol design due to its volume capacity and length It is compatible with all 96 multi-channel heads on Biomek i-Series (i5 and i7) NX/FX/FXP and Multimek Alkali Scientificmanufactures its P250 black conductive tips from premium virgin polypropylene and utilizes advanced polishing techniques to ensure minimal liquid retention without using Specifications Working volume 190 00 L Well format 96 96 384 plate accessible Head compatibility 96 multi-channel Beckman