Learn More
Alkali Scientific 190uL Beckman Biomek FX/NX Mul
SDP
Catalog No. 503026945
Encompass_Preferred
Alkali Scientificdesigns its P250 automation tips for optimal compatibility with Beckman Coulter Biomek i-Series (i5 and i7) as well as previous NX/FX/FXP robotic liquid handlers Using Alkali Scientificautomation tips eliminates the need for a separate labware definition and ensures that existing automation control software protocols remain unchanged The Alkali ScientificP250 (250 L) tip is among the most widely used in the 96-format category offering excellent precision and accuracy for sub-microliter volumes Its capacity and length make it versatile for various protocol designs These tips are compatible with all 96 multi-channel heads on Biomek i-Series (i5 and i7) NX/FX/FXP and Multimek systems Manufactured from premium-grade virgin polypropylene Alkali Scientificensures minimal liquid retention in the P250 tips through advanced polishing techniques without the use of releasing agents or additives Specifications Working volume 190 00 L Well format 96 96 384 plate accessible
Catalog No. 50-302-6945 Supplier Alkali Scientific Supplier No. FXS180CS
Alkali Scientificdesigns its P250 automation tips for optimal compatibility with Beckman Coulter Biomek i-Series (i5 and i7) as well as previous NX/FX/FXP robotic liquid handlers Using Alkali Scientificautomation tips eliminates the need for a separate labware definition and ensures that existing automation control software protocols remain unchanged The Alkali ScientificP250 (250 L) tip is among the most widely used in the 96-format category offering excellent precision and accuracy for sub-microliter volumes Its capacity and length make it versatile for various protocol designs These tips are compatible with all 96 multi-channel heads on Biomek i-Series (i5 and i7) NX/FX/FXP and Multimek systems Manufactured from premium-grade virgin polypropylene Alkali Scientificensures minimal liquid retention in the P250 tips through advanced polishing techniques without the use of releasing agents or additives Specifications Working volume 190 00 L Well format 96 96 384 plate accessible