Learn More
Ace Glass, Inc. 190x100mm crystallization dish, cs/6, sp/2, 3140-190
SDP
Catalog No. 509558484
Encompass
190X100mm Corning crystallization dish made of PYREX glass. Rims are reinforced and fire polished to reduce chipping.
This item is not returnable. View return policy
Catalog No. 50-955-8484 Supplier Ace Glass, Inc. Supplier No. 846522
190X100mm Corning crystallization dish made of PYREX glass. Rims are reinforced and fire polished to reduce chipping.