Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Sure-Grip™ EX Flammable Safety Cabinet
GREENER_CHOICE

Catalog No. 191202338 Shop All Justrite Safety Group Products
Change view
Click to view available options
Load Bearing Capacity:
30 gal.
45 gal.
60 gal.
90 gal.
Dimensions (D x W x H) Interior:
14.56 x 39.5 x 39.62 in.
14.56 x 39.5 x 60.5 in.
30.34 x 30.75 x 60.5 in.
30.34 x 39.5 x 60.5 in.
Door Style:
Manual-close
Self-close
Weight (Metric):
105 kg
108 kg
141 kg
142 kg
176 kg
218 kg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Load Bearing Capacity Dimensions (D x W x H) Interior Door Style Weight (Metric)
19-120-2338 60 gal. 30.34 x 30.75 x 60.5 in. Self-close 218 kg
19-120-2333 30 gal. 14.56 x 39.5 x 39.62 in. Manual-close 105 kg
19-120-2334 30 gal. 14.56 x 39.5 x 39.62 in. Self-close 176 kg
19-120-2335 45 gal. 14.56 x 39.5 x 60.5 in. Manual-close 108 kg
19-120-2339 90 gal. 30.34 x 39.5 x 60.5 in. Manual-close 142 kg
19-120-2340 90 gal. 30.34 x 39.5 x 60.5 in. Self-close 218 kg
19-120-2337 60 gal. 30.34 x 30.75 x 60.5 in. Manual-close 141 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-120-2338 Supplier Justrite™ Supplier No. 896020
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.

Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.

  • Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
  • Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
  • Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
  • Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
  • Self-latch engages critical three-point latching system for protection under fire conditions
  • Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
  • Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
  • Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
  • SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
  • Adjustable leveling feet ensure solid positioning and alignment for tight door closure
  • Exterior side panel has built-in grounding connector for easy grounding
  • Finished inside and out with durable lead-free powder paint
  • Meet OSHA and NFPA requirements

Certifications

FM approved except 19-120-2336.

Compliance

Meet NFPA Code 30 and OSHA regulations; models with self-close doors also comply with the Uniform Fire Code.

Warranty and Services

10-Year

Specifications

Dimensions (D x W x H) Interior 30.34 x 30.75 x 60.5 in.
Product Type Can Storage Cabinet
Load Bearing Capacity 60 gal.
Door Style Self-close
No. of Doors 2
Width (Metric) Interior 78 cm
Width (Metric) Exterior 86.4 cm
No. of Shelves 2
Height (Metric) Exterior 165.1 cm
Depth (Metric) Exterior 86.4 cm
Depth (Metric) Interior 77.1 cm
Height (Metric) Interior 153.7 cm
Weight (Metric) 218 kg
Surface Coating Epoxy, Polyester
Certifications/Compliance FM, NFPA, OSHA, U (NFPA)
Weight (English) 480 lb.
Color Yellow
Material Steel (CR)
Includes U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with
Depth (English) Exterior 34 in.
Width (English) Interior 30.75 in.
For Use With (Application) Flammable liquid storage and protection
Width (English) Exterior 34 in.
Depth (English) Interior 30.34 in.
Height (English) Exterior 65 in.
Height (English) Interior 60.5 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.