Learn More
Pyramex™ Rendezvous™ Safety Glasses
Catalog No. 19152341 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
Blue
Crystal Gray
Lens Coating:
Scratch-resistant
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor Mirror
Infinity Blue
Silver Mirror
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
19-152-341 Crystal Gray Scratch-resistant Clear
19-121-004 Black Scratch-resistant Clear
19-152-340 Black Scratch-resistant Clear
19-121-005 Black - Gray
19-152-343 Black - Amber
19-121-009 Black Scratch-resistant Indoor/Outdoor Mirror
19-121-006 Black - Infinity Blue
19-121-008 Black - Gray
19-121-007 Black - Silver Mirror
19-121-010 Blue - Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-152-341 Supplier Pyramex™ Supplier No. SCG2810ST
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Adjustable lens angle aligns to the proper position for various work applications
Ratcheting lens pitch adjustment aligns lens angle to properly position for work application.
  • Flexible nylon temples with four size settings to accommodate a broad range of facial sizes
  • Adjustable nose pad conforms to a broad range of bridge sizes
  • Lightweight nylon frame for comfortable, all day wear
  • Replaceable, 9.5 base-curved polycarbonate lens withstands high-impact
  • Provide 99% UV protection, is completely distortion free
Compliance
Meet ANSI Z87.1 requirements.
Specifications
Certifications/Compliance Meet ANSI Z87.1 requirements.
Lens Coating Scratch-resistant
Lens Tint Clear
For Use With (Application) For indoor applications
Frame Color Crystal Gray
Lens Material Polycarbonate
Frame Material Polycarbonate