Learn More
eMolecules 1952360-91-6 | (Fmoc-amino)-PEG12-C2-carboxylic acid | Chem-Impex | MFCD08064302 | 839.973 | C42H65NO16 | 98.000 | OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(=O)OCC1c2ccccc2-c2ccccc12 | 5g | 686069703
Shop All eMolecules Inc. Products