Learn More
eMolecules​ 19814-75-6 | 9,9-DIMETHYL-9H-XANTHENE | AstaTech | MFCD00134434 | 210.276 | C15H14O | 95.000 | CC1(C)c2ccccc2Oc2ccccc12 | 500g | 411870555
Catalog No. 5000491466 Shop All eMolecules Inc. Products
Encompass_Preferred
9,9-DIMETHYL-9H-XANTHENE | AstaTech | 19814-75-6 | MFCD00134434 | 210.276 | C15H14O | 95.000 | CC1(C)c2ccccc2Oc2ccccc12 | 500g | 411870555
Catalog No. 50-004-91466 Supplier eMolecules​ Supplier No. 411870555
9,9-DIMETHYL-9H-XANTHENE | AstaTech | 19814-75-6 | MFCD00134434 | 210.276 | C15H14O | 95.000 | CC1(C)c2ccccc2Oc2ccccc12 | 500g | 411870555