Learn More
eMolecules​ 199593-09-4 | 2'-OMe-Ac-C Phosphoramidite | ChemScene | MFCD12912403 | 801.878 | C42H52N5O9P | 97.000 | CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c2ccc(OC)cc2)c2ccc(OC)cc2)O[C@H]1n1ccc(NC(C)=O)nc1=O | 10g | 687111539
Catalog No. NC3930488 Shop All eMolecules Inc. Products
Encompass_Preferred
2'-OMe-Ac-C Phosphoramidite | ChemScene | 199593-09-4 | MFCD12912403 | 801.878 | C42H52N5O9P | 97.000 | CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c2ccc(OC)cc2)c2ccc(OC)cc2)O[C@H]1n1ccc(NC(C)=O)nc1=O | 10g | 687111539
Catalog No. NC3930488 Supplier eMolecules​ Supplier No. 687111539
2'-OMe-Ac-C Phosphoramidite | ChemScene | 199593-09-4 | MFCD12912403 | 801.878 | C42H52N5O9P | 97.000 | CO[C@@H]1[C@H](OP(OCCC#N)N(C(C)C)C(C)C)[C@@H](COC(c2ccccc2)(c2ccc(OC)cc2)c2ccc(OC)cc2)O[C@H]1n1ccc(NC(C)=O)nc1=O | 10g | 687111539