Learn More
Ace Glass, Inc. 19L solution bottle plastic coated
SDP
Catalog No. 50971858
Encompass_Preferred
19L solution bottle is coated to the neck with PVC safety coating to reduce potential breakage, and exposure to laboratory personnel when handling hazardous material. Carboy style bottles with sloping shoulders. Necks are tooled for uniform fit with No. 12 stoppers (not supplied).
Catalog No. 50-971-858 Supplier Ace Glass, Inc. Supplier No. 539309
19L solution bottle is coated to the neck with PVC safety coating to reduce potential breakage, and exposure to laboratory personnel when handling hazardous material. Carboy style bottles with sloping shoulders. Necks are tooled for uniform fit with No. 12 stoppers (not supplied).