Learn More
Ace Glass, Inc. 19mm quartz graded seals, length 7.6cm
SDP
Supplier:  Ace Glass, Inc. 845530
Encompass
19mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-600