Learn More
Chemglass Life Sciences Chemglass 19 mm Mechanical Seal Bearing Repair Kit (for CG-2077-M series)
SDP
Catalog No. 502733535
Encompass_Preferred
Repair kit containing replacement internal components to refurbish 19 mm mechanical seal stir bearings. Supplied without tools; grease is not included and must be purchased separately.
  • Includes bearing seal, ball bearings, and needle roller bearings.
  • Services 19 mm mechanical seal stir bearings.
  • Sold without tools for field installation or maintenance.
  • Requires a small amount of high-vacuum grease, sold separately.
Catalog No. 50-273-3535 Supplier Chemglass Life Sciences Supplier No. CG2077M319
Repair kit containing replacement internal components to refurbish 19 mm mechanical seal stir bearings. Supplied without tools; grease is not included and must be purchased separately.
  • Includes bearing seal, ball bearings, and needle roller bearings.
  • Services 19 mm mechanical seal stir bearings.
  • Sold without tools for field installation or maintenance.
  • Requires a small amount of high-vacuum grease, sold separately.