Learn More
Chemglass Life Sciences 19 mm Adjustable Probe Holder, G3/4 Thread - For Bioreactors
SDP
Catalog No. NC2892877
Encompass
This 19 mm adjustable-height probe holder provides a sealed, corrosion-resistant mounting for process probes in laboratory and pilot-scale bioreactors. It features a G3/4 threaded connection and a compression fitting for precise probe positioning.
  • Adjustable height using a compression fitting for precise probe placement.
  • Accepts 19 mm diameter probes.
  • G3/4 threaded connection for secure installation.
  • 316L stainless steel construction with electropolished surface for corrosion resistance.
  • Suitable for use as a replacement probe holder in laboratory bioreactors.
Catalog No. NC2892877 Supplier Chemglass Life Sciences Supplier No. CLS138110
This 19 mm adjustable-height probe holder provides a sealed, corrosion-resistant mounting for process probes in laboratory and pilot-scale bioreactors. It features a G3/4 threaded connection and a compression fitting for precise probe positioning.
  • Adjustable height using a compression fitting for precise probe placement.
  • Accepts 19 mm diameter probes.
  • G3/4 threaded connection for secure installation.
  • 316L stainless steel construction with electropolished surface for corrosion resistance.
  • Suitable for use as a replacement probe holder in laboratory bioreactors.