Learn More
Chemglass Life Sciences 19mm PTFE Gasket
SDP
Catalog No. CG204902
Encompass_Preferred
19mm PTFE Gasket. PTFE gasket is 0.015" thick with an opening to fit Chemglass stirrer shafts. Gasket is placed below the CG-2048 Safety collar to prevent wear as a result of the collar rotating against the stirrer bearing. Also useful as a dust cover. SOLD 12/PKG.
Catalog No. CG204902 Supplier Chemglass Life Sciences Supplier No. CG204902
19mm PTFE Gasket. PTFE gasket is 0.015" thick with an opening to fit Chemglass stirrer shafts. Gasket is placed below the CG-2048 Safety collar to prevent wear as a result of the collar rotating against the stirrer bearing. Also useful as a dust cover. SOLD 12/PKG.