Learn More
Chemglass Life Sciences Chemglass 19 mm Rotary Bearing Rebuild Kit with Tools - for CG-2077-M series mechanical-seal stirrer bearings
SDP
Catalog No. NC2745506
Encompass_Preferred
The Chemglass 19 mm rotary bearing rebuild kit supplies replacement bearings, seals, and hand tools needed to service 19 mm mechanical-seal stirrer bearings. It supports routine maintenance and seal replacement for rounded or beveled glass, metal, or PTFE-covered stir shafts.
  • Includes adjustable pin spanner wrench and retaining ring pliers.
  • Contains bearing seal, ball bearings, and needle roller bearings.
  • Bearing body material: glass-filled PTFE.
  • Seals use perfluorinated O-rings for broad chemical resistance.
  • Rated seals up to 2,000 rpm; manufacturer recommends a maximum of 500 rpm with glass vessels or shafts.
  • Seal life approximately 3,000 hours.
  • Specialized grease sold separately; a small amount of mineral oil may be applied during installation.
  • Designed for use with rounded or beveled stir shafts; do not use with flat-ended shafts.
Catalog No. NC2745506 Supplier Chemglass Life Sciences Supplier No. CG2077M219
The Chemglass 19 mm rotary bearing rebuild kit supplies replacement bearings, seals, and hand tools needed to service 19 mm mechanical-seal stirrer bearings. It supports routine maintenance and seal replacement for rounded or beveled glass, metal, or PTFE-covered stir shafts.
  • Includes adjustable pin spanner wrench and retaining ring pliers.
  • Contains bearing seal, ball bearings, and needle roller bearings.
  • Bearing body material: glass-filled PTFE.
  • Seals use perfluorinated O-rings for broad chemical resistance.
  • Rated seals up to 2,000 rpm; manufacturer recommends a maximum of 500 rpm with glass vessels or shafts.
  • Seal life approximately 3,000 hours.
  • Specialized grease sold separately; a small amount of mineral oil may be applied during installation.
  • Designed for use with rounded or beveled stir shafts; do not use with flat-ended shafts.