Learn More
Chemglass Life Sciences 19mm Rotary Bearing Taper Lock, Ferrule, Nylon
SDP
Catalog No. 501531432
Encompass_Preferred
19mm Rotary Bearing Taper Lock, Ferrule, Nylon. Component of CG-2077-M series of mechanical seal stir bearings. Pictured is a fully assembled bearing. Item is ROTARY BEARING TAPER LOCK FERRULE ONLY.
Catalog No. 50-153-1432 Supplier Chemglass Life Sciences Supplier No. CG2077M119
19mm Rotary Bearing Taper Lock, Ferrule, Nylon. Component of CG-2077-M series of mechanical seal stir bearings. Pictured is a fully assembled bearing. Item is ROTARY BEARING TAPER LOCK FERRULE ONLY.