Learn More
Chemglass Life Sciences Replacement 19mm Stirrer Shaft, Reactor Size: 10L/15L/20L Unjacketed, Fits Support Frame: CG-1965-X-150, Overall Length: 29-3/16"
SDP
Catalog No. 50872424
Encompass_Preferred
Replacement 19mm Stirrer Shaft, Reactor Size: 10L/15L/20L Unjacketed, Fits Support Frame: CG-1965-X-150, Overall Length: 29-3/16". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. 50-872-424 Supplier Chemglass Life Sciences Supplier No. CG209711
Replacement 19mm Stirrer Shaft, Reactor Size: 10L/15L/20L Unjacketed, Fits Support Frame: CG-1965-X-150, Overall Length: 29-3/16". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.