Learn More
Chemglass Life Sciences Replacement 19mm Stirrer Shaft, Reactor Size: 15L-20L, Fits Support Frame: CG-1965-X-150, Overall Length: 34.6"
SDP
Catalog No. 50872526
Encompass_Preferred
Replacement 19mm Stirrer Shaft, Reactor Size: 15L-20L, Fits Support Frame: CG-1965-X-150, Overall Length: 34.6". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat. REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.
Catalog No. 50-872-526 Supplier Chemglass Life Sciences Supplier No. CG209712
Replacement 19mm Stirrer Shaft, Reactor Size: 15L-20L, Fits Support Frame: CG-1965-X-150, Overall Length: 34.6". Precision ground and polished stirrer shaft for use with Chemglass process reactors. Designed to utilize CG-2095 and CG-2096 PTFE agitators. Lower portion of stirrer shaft is sandblasted and has a 3/8¿ wide flat. REPLACEMENT STIRRER SHAFTS ARE FOR USE WITH MECHANICAL SEAL STIR BEARING ON LISTED SUPPORT FRAME.