Learn More
Chemglass Life Sciences Stirrer Blade, PTFE, Banana Shaped, Solid, 19mm, Heavy Duty, for 3000mL Flasks, 34/45 minimum joint clearance
SDP
Catalog No. 50872520
Encompass_Preferred
Stirrer Blade, PTFE, Banana Shaped, Solid, 19mm, Heavy Duty, for 3000mL Flasks, 34/45 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are solid. Made of 3/16" PTFE to fit 19mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.
Catalog No. 50-872-520 Supplier Chemglass Life Sciences Supplier No. CG209003
Stirrer Blade, PTFE, Banana Shaped, Solid, 19mm, Heavy Duty, for 3000mL Flasks, 34/45 minimum joint clearance. Blade is shaped to resemble the contour of a round bottom flask. Blades are solid. Made of 3/16" PTFE to fit 19mm OD stirrer shafts. Minimum joint size listed is the size required to allow passage of the blade.