Learn More
Chemglass Life Sciences Stirring Shaft, 19mm, Polished, Button Style, for 12000 to 20000mL Flasks, 725mm OAL
SDP
Catalog No. CG208802
Encompass_Preferred
Stirring Shaft, 19mm, Polished, Button Style, for 12000mL to 22000mL Flasks, 725mm OAL. A 19mm precision ground and polished stirrer shaft to fit CG-2086 19mm PTFE bearings. Side button for attachment of CG-2089 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2085.ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.
Catalog No. CG208802 Supplier Chemglass Life Sciences Supplier No. CG208802
Stirring Shaft, 19mm, Polished, Button Style, for 12000mL to 22000mL Flasks, 725mm OAL. A 19mm precision ground and polished stirrer shaft to fit CG-2086 19mm PTFE bearings. Side button for attachment of CG-2089 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2085.ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.