Learn More
Chemglass Life Sciences Stirring Shaft, 19mm, Polished, Button Style, for 50000 to 72000mL Flasks, 950mm OAL
SDP
Catalog No. CG208803
Encompass_Preferred
Stirring Shaft, 19mm, Polished, Button Style, for 50000mL to 72000mL Flasks, 950mm OAL. A 19mm precision ground and polished stirrer shaft to fit CG-2086 19mm PTFE bearings. Side button for attachment of CG-2089 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2085.ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.
Catalog No. CG208803 Supplier Chemglass Life Sciences Supplier No. CG208803
Stirring Shaft, 19mm, Polished, Button Style, for 50000mL to 72000mL Flasks, 950mm OAL. A 19mm precision ground and polished stirrer shaft to fit CG-2086 19mm PTFE bearings. Side button for attachment of CG-2089 PTFE stirrer blades.PLEASE NOTE: THESE SHAFTS ARE NOT TO BE USED WITH ANY TYPE OF GLASS BEARING. FOR GROUND SHAFTS USED WITH GLASS BEARINGS SEE CG-2085.ALSO NOTE: 19MM SHAFTS HAVE A REDUCED DIAMETER TOP SECTION THAT IS 1/2"OD, PLEASE KEEP THIS IN MIND WHEN SELECTING THE PROPER SIZE COUPLING.