Learn More
Ace Glass, Inc. 19mm x 25cm chromatography column
SDP
Supplier:  Ace Glass, Inc. 588825
Encompass_Preferred
19mm x 25cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Catalog No. 50-972-546