Learn More
Ace Glass, Inc. 19mm x 51cm chromatography column
SDP
Catalog No. 50972547
Encompass_Preferred
19mm x 51cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.
Catalog No. 50-972-547 Supplier Ace Glass, Inc. Supplier No. 588835
19mm x 51cm chromatography column with 2mm bore 1:5 taper solid PTFE stopcock plug at the bottom.