Learn More
Ace Glass, Inc. 19mm x 61cm chromatography column
SDP
Catalog No. 50972558
Encompass_Preferred
19mm x 61cm chromatography column with #15 Ace-Thred at top and 2mm bore 1:5 taper solid PTFE stopcock plug at bottom. Column supplied with ACE 7506-06 nylon bushing and FETFE o-ring.
Catalog No. 50-972-558 Supplier Ace Glass, Inc. Supplier No. 590221
19mm x 61cm chromatography column with #15 Ace-Thred at top and 2mm bore 1:5 taper solid PTFE stopcock plug at bottom. Column supplied with ACE 7506-06 nylon bushing and FETFE o-ring.