Learn More
CLARE CHEMICAL RESEARCH 19 x 15 cm Dark Reader Mid-size Blue Light LED Transilluminator
Supplier:  CLARE CHEMICAL RESEARCH DR46B
Encompass
Our mid-size blue light LED transilluminator is designed to safely view DNA bands in agarose gels without any DNA damage. Enhance cloning efficiencies up to 1000-fold! Excels with the newest fluorescent DNA stains. Viewing glasses included.
Catalog No. NC9800797