Learn More
TARGETMOL CHEMICALS INC 1A-116 5MG
SDP
Catalog No. 502878692
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.1A-116 is a specific Rac1 inhibitor. Purity 98.43%
Catalog No. 50-287-8692 Supplier TARGETMOL CHEMICALS INC Supplier No. T140045MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.1A-116 is a specific Rac1 inhibitor. Purity 98.43%