Learn More
Enzo Life Sciences 1alpha,25-Dihydroxyvitamin D3 (1mg). CAS: 32222-06-3
SDP
Supplier:  Enzo Life Sciences BMLDM2001000
Encompass_Preferred
1α,25-Dihydroxyvitamin D3 is the active hormonal form of vitamin D. It inhibits proliferation and induces differentiation of a variety of cells including osteoclasts, HL-60 cells. It is a central regulator of bone and calcium homeostasis. A polymorphism in the vitamin D receptor gene has been linked to a genetic predisposition to osteoporosis. It inhibits proliferation and induces differentiation of a variety of cells including osteoclasts, keratinocytes, HL-60 cells vascular smooth muscle cells and bone marrow macrophage precursors. Alternative name: 1alpha,25-Dihydroxycholecalciferol, Calcitriol. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO, 100% ethanol,  ethyl acetate, tetrahydrofuran, chloroform or acetone. Evaporative loss of solvent may occur during storage and handling of the dissolved product. If necessary, exact concentration may be redetermined spectrophotometrically. Long Term Storage: -80°C.
Catalog No. 50-201-0516