Learn More
ALADDIN SCIENTIFIC CORPORATION 1a1-Diphenyl-2-picrylhydrazyl , 100mg
Catalog No. 5000082567
Encompass_Preferred
22-Diphenyl-1-picrylhydrazyl is used as a sensitive colorimetric free radical scavenger It serves as a general antioxidant detector It is utilized to monitor chemical reactions involving radicals as well as a standard of the position and intensity of electron paramagnetic resonance signals Further it is employed in the quantitative determination of aliphatic and aromatic thiols by indirect spectroscopy and for photometric determination of tocopherol
Catalog No. 50-000-82567 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. D273092100MG
22-Diphenyl-1-picrylhydrazyl is used as a sensitive colorimetric free radical scavenger It serves as a general antioxidant detector It is utilized to monitor chemical reactions involving radicals as well as a standard of the position and intensity of electron paramagnetic resonance signals Further it is employed in the quantitative determination of aliphatic and aromatic thiols by indirect spectroscopy and for photometric determination of tocopherol