Learn More
ALADDIN SCIENTIFIC CORPORATION 1a3a5-Trimethylbenzene, 100ml
Catalog No. 5000082514
Encompass_Preferred
Mesitylene may be used to-Grow fullerene(C60)/135-trimethylbenzene (mesitylene) nanowires Prepare 2 4 6 -trimethyl acetophenone with high slectivity by reacting with acetyl chloride in the presence of dodecatungstophosphoric acid (DTP)/K-10 montomorillonite clay Synthesize 35-dimethylbenzaldehyde from benzene via peroxidative oxidation in the presence of vanadium(IV or V) compounds with NO-ligands as catalysts
Catalog No. 50-000-82514 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. T139982100ML
Mesitylene may be used to-Grow fullerene(C60)/135-trimethylbenzene (mesitylene) nanowires Prepare 2 4 6 -trimethyl acetophenone with high slectivity by reacting with acetyl chloride in the presence of dodecatungstophosphoric acid (DTP)/K-10 montomorillonite clay Synthesize 35-dimethylbenzaldehyde from benzene via peroxidative oxidation in the presence of vanadium(IV or V) compounds with NO-ligands as catalysts