Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1alpha, 24, 25-Trihydroxy VD2 | 457048-34-9 | C28H44O4 | 1 MG
SDP

Catalog No. 5000372170
Encompass_Preferred

1alpha, 24, 25-Trihydroxy VD2 is a vitamin D analog.

  • Purity of 98.21%
  • Molecular weight of 444.65
  • Appears as a white to yellow solid
  • For research use only
  • Optimal storage at -20°C, protected from light and stored under nitrogen
  • Soluble in DMSO at 12.5 mg/mL

Catalog No. 50-003-72170 Supplier Medchemexpress LLC Supplier No. HY151561MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1alpha, 24, 25-Trihydroxy VD2 is a vitamin D analog.

  • Purity of 98.21%
  • Molecular weight of 444.65
  • Appears as a white to yellow solid
  • For research use only
  • Optimal storage at -20°C, protected from light and stored under nitrogen
  • Soluble in DMSO at 12.5 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.