Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarbonitrile 98.0+%, TCI America™
SDP

Catalog No. C19671G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C19671G Supplier TCI America Supplier No. C19671G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 168280-46-4
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
InChI Key VMUOSHREZKXCIV-UHFFFAOYSA-N
Synonym (1alpha,3alpha,5alpha)-1,3,5-Tricyanocyclohexane
PubChem CID 11423741
IUPAC Name cyclohexane-1,3,5-tricarbonitrile
SMILES C1C(CC(CC1C#N)C#N)C#N

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 159.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1alpha,3alpha,5alpha)-1,3,5-Cyclohexanetricarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.