Learn More
Cayman Chemical 1AmIno1deoxyDfructoseh 100mg
SDP
Catalog No. 502991775
Encompass_Preferred
An amino monosaccharide; induces site-specific DNA damage at pyrimidine residues in cell-free assays; 1-amino-1-deoxy-D-fructose moieties have been found as a component of fructosamines
Catalog No. 50-299-1775 Supplier Cayman Chemical Supplier No. 30571100 MG
An amino monosaccharide; induces site-specific DNA damage at pyrimidine residues in cell-free assays; 1-amino-1-deoxy-D-fructose moieties have been found as a component of fructosamines