Learn More
Cayman Chemical 1-Amino-1-deoxy-D-fructose (hydrochloride), Purity: ≥95%, CAS: 39002-30-7, 50 mg
SDP
Catalog No. 502991296
Encompass_Preferred
An amino monosaccharide; induces site-specific DNA damage at pyrimidine residues in cell-free assays; 1-amino-1-deoxy-D-fructose moieties have been found as a component of fructosamines
Catalog No. 50-299-1296 Supplier Cayman Chemical Supplier No. 3057150 MG
An amino monosaccharide; induces site-specific DNA damage at pyrimidine residues in cell-free assays; 1-amino-1-deoxy-D-fructose moieties have been found as a component of fructosamines