Learn More
Cayman Chemical 1-Aminodecylidene bis-Phosphonic Acid, Purity: ≥95%, CAS: 15049-93-1, 5 mg
SDP
Catalog No. 502955106
Encompass_Preferred
A potent inhibitor of acid sphingomyelinase (IC50 = 20 nM); selective for acid sphingomyelinase over neutral sphingomyelinase (IC50 = >100 μM); inhibits dexamethasone-induced apoptosis in HepG2 cells
Catalog No. 50-295-5106 Supplier Cayman Chemical Supplier No. 135835 MG
A potent inhibitor of acid sphingomyelinase (IC50 = 20 nM); selective for acid sphingomyelinase over neutral sphingomyelinase (IC50 = >100 μM); inhibits dexamethasone-induced apoptosis in HepG2 cells