Learn More
Cayman Chemical 1-Arachidoyl-2-hydroxy-sn-glycero-3-PC, Purity: ≥95%, CAS: 108341-80-6, 10 mg
SDP
Catalog No. 502916658
Encompass_Preferred
A lysophospholipid; plasma levels are increased in ApoE-/- mice fed a high-fat diet
Catalog No. 50-291-6658 Supplier Cayman Chemical Supplier No. 3830310 MG
A lysophospholipid; plasma levels are increased in ApoE-/- mice fed a high-fat diet