Learn More
Cole-Parmer Ashcroft 1032S2.0 2" Dry Sanitary Gauge 30" Hg to 100 psi, 3/4 Tri-Clamp
Catalog No. NC1556431 Shop All Cole-Parmer Products
Encompass_Preferred
The Ashcroft Type 1032 Sanitary Pressure Gauge is specifically designed for applications in the food, pharmaceutical and biotechnical industries where small size and sanitary conditions are a priority. This compact autoclavable 2" gauge features all stainless steel construction, temperature-vented case, built-in pressure dampening and a self-draining case to facilitate wash downs. The gauges can also be cleaned or steamed in place.
Catalog No. NC1556431 Supplier Cole-Parmer Supplier No. 6846042
The Ashcroft Type 1032 Sanitary Pressure Gauge is specifically designed for applications in the food, pharmaceutical and biotechnical industries where small size and sanitary conditions are a priority. This compact autoclavable 2" gauge features all stainless steel construction, temperature-vented case, built-in pressure dampening and a self-draining case to facilitate wash downs. The gauges can also be cleaned or steamed in place.